Learn More
Biomatik Corporation Pig Haptoglobin (Hpt) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04636
Encompass_Preferred
Pig Haptoglobin (Hpt) ELISA Kit for detecting Hpt in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.2-2,000ng/ml. Sensitivity: 15.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8SPS7. Gene ID: 397061. Target Synonyms: HP; Hp2-Alpha; Alpha-2-Macroglobulin; Zonulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2683