Learn More
Biomatik Corporation Mouse Cluster Of Differentiation 8a (CD8a) ELISA Kit
Supplier:  Biomatik Corporation EKU09045
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P01731. Target Synonyms: p32; CD8-A; Leu2; MAL; T-cell surface glycoprotein CD8 alpha chain; T-lymphocyte differentiation antigen T8/Leu-2. Restriction: For Research Use Only.
Catalog No. 50-228-6570