Learn More
Biomatik Corporation Rat Haptoglobin (Hpt) ELISA Kit, 96 tests
Catalog No. 501502684
Encompass_Preferred
Rat Haptoglobin (Hpt) ELISA Kit for detecting Hpt in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000ng/ml. Sensitivity: 27.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06866. Gene ID: 24464. Target Synonyms: HP; Hp2-Alpha; Alpha-2-Macroglobulin; Zonulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2684 Supplier Biomatik Corporation Supplier No. EKU04637
Rat Haptoglobin (Hpt) ELISA Kit for detecting Hpt in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000ng/ml. Sensitivity: 27.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06866. Gene ID: 24464. Target Synonyms: HP; Hp2-Alpha; Alpha-2-Macroglobulin; Zonulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.