Learn More
Biomatik Corporation Human Carnitine Acetyltransferase (CRAT) ELISA Kit
Catalog No. 502286579
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 3.12-200ng/mL. Sensitivity: 1.14ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P43155. Target Synonyms: CAT1; Carnitine O-Acetyltransferase; Carnitine acetylase. Restriction: For Research Use Only.
This item is not returnable. View return policy
Catalog No. 50-228-6579 Supplier Biomatik Corporation Supplier No. EKU09118
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 3.12-200ng/mL. Sensitivity: 1.14ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P43155. Target Synonyms: CAT1; Carnitine O-Acetyltransferase; Carnitine acetylase. Restriction: For Research Use Only.