Learn More
Biomatik Corporation Human Heat Shock 27kDa Protein 1 (HSPB1) ELISA Kit, 96 tests
Catalog No. 501502685
Encompass_Preferred
The Human Heat Shock 27kDa Protein 1 (HSPB1) ELISA Kit allows for quantitative determination of Heat Shock 27kDa Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.33 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: p04792. Cross Activity: No significant cross-reactivity or interference between Heat Shock 27kDa Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2685 Supplier Biomatik Corporation Supplier No. EKU04643
The Human Heat Shock 27kDa Protein 1 (HSPB1) ELISA Kit allows for quantitative determination of Heat Shock 27kDa Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.33 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: p04792. Cross Activity: No significant cross-reactivity or interference between Heat Shock 27kDa Protein 1 and analogues was observed. Restriction: For Research Use Only.