Learn More
Biomatik Corporation Mouse Heat Shock 27kDa Protein 1 (HSPB1) ELISA Kit, 96 tests
Catalog No. 501502686
Encompass_Preferred
Mouse Heat Shock Protein 27 (Hsp27) ELISA Kit for detecting Hsp27 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14602. Gene ID: 15507. Target Synonyms: HSPB1; HSP-B1; CMT2F; HSP28; Hsp25; Heat Shock 27kDa Protein 1; 28 kDa heat shock protein; Heat shock protein beta-1; Estrogen-regulated 24 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2686 Supplier Biomatik Corporation Supplier No. EKU04644
Mouse Heat Shock Protein 27 (Hsp27) ELISA Kit for detecting Hsp27 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14602. Gene ID: 15507. Target Synonyms: HSPB1; HSP-B1; CMT2F; HSP28; Hsp25; Heat Shock 27kDa Protein 1; 28 kDa heat shock protein; Heat shock protein beta-1; Estrogen-regulated 24 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.