Learn More
Biomatik Corporation Human Heat Shock 70kDa Protein 1A (HSPA1A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04654
Encompass_Preferred
The Human Heat Shock 70kDa Protein 1A (HSPA1A) ELISA Kit allows for quantitative determination of Heat Shock 70kDa Protein 1A in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.60 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08107. Cross Activity: No significant cross-reactivity or interference between Heat Shock 70kDa Protein 1A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2690