Learn More
Biomatik Corporation Human Heat Shock 70kDa Protein 2 (HSPA2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04659
Encompass_Preferred
The Human Heat Shock 70kDa Protein 2 (HSPA2) ELISA Kit allows for quantitative determination of Heat Shock 70kDa Protein 2 in Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.69 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P54652. Cross Activity: No significant cross-reactivity or interference between Heat Shock 70kDa Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2691