Learn More
Biomatik Corporation Human Heat Shock 70kDa Protein 8 (HSPA8) ELISA Kit, 96 tests
Catalog No. 501502693
Encompass_Preferred
The Human Heat Shock 70kDa Protein 8 (HSPA8) ELISA Kit allows for quantitative determination of Heat Shock 70kDa Protein 8 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11142. Cross Activity: No significant cross-reactivity or interference between Heat Shock 70kDa Protein 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2693 Supplier Biomatik Corporation Supplier No. EKU04663
The Human Heat Shock 70kDa Protein 8 (HSPA8) ELISA Kit allows for quantitative determination of Heat Shock 70kDa Protein 8 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11142. Cross Activity: No significant cross-reactivity or interference between Heat Shock 70kDa Protein 8 and analogues was observed. Restriction: For Research Use Only.