Learn More
Biomatik Corporation Mouse Heat Shock Protein 40 (HSP40) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04667
Encompass_Preferred
Mouse Heat Shock Protein 40 (HSP40) ELISA Kit for detecting HSP40 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.242ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QYJ3. Gene ID: 81489. Target Synonyms: DnaJ; DNAJB1; Sis1; RSPH16B; HSPF1; DnaJ homolog subfamily B member 1; Radial Spoke 16 Homolog B; Heat Shock 40kDa Protein 1; DnaJ protein homolog 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2696