Learn More
Biomatik Corporation Human Leucine Rich Repeat Kinase 2 (LRRK2) ELISA Kit
Supplier:  Biomatik Corporation EKU11041
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 78-5,000pg/mL. Sensitivity: 31pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q5S007. Target Synonyms: AURA17; PARK8; RIPK7; ROCO2; Dardarin ; Parkinson Disease(Autosomal Dominant)8; Leucine-rich repeat serine/threonine-protein kinase 2. Restriction: For Research Use Only.
Catalog No. 50-228-6844