Learn More
Biomatik Corporation Human Procollagen II C-Terminal Propeptide (PIICP) ELISA Kit
Supplier:  Biomatik Corporation EKU11360
Encompass_Preferred
Human Procollagen II C-Terminal Propeptide (PIICP) ELISA Kit for detecting PIICP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.47-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02458. Gene ID: 1280. Target Synonyms: P2CP; C-Propeptide Of Type II Procollagen; Procollagen II Carboxy Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6903