Learn More
Biomatik Corporation Human Signal Transducer And Activator Of Transcription 3 (STAT3) ELISA Kit
Supplier:  Biomatik Corporation EKU11428
Encompass_Preferred
Human Signal Transducer And Activator Of Transcription 3 (STAT3) ELISA Kit for detecting STAT3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40763. Gene ID: 6774. Target Synonyms: APRF; Acute-Phase Response Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6918