Learn More
Biomatik Corporation Mouse Heme Oxygenase 1, Decycling (HO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04707
Encompass_Preferred
Mouse Heme Oxygenase 1 (HO1) ELISA Kit for detecting HO1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14901. Gene ID: 15368. Target Synonyms: HMOX1; Hsp32; HMOX1D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2709