Learn More
Biomatik Corporation Rat Heme Oxygenase 1, Decycling (HO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04708
Encompass_Preferred
Rat Heme Oxygenase 1 (HO1) ELISA Kit for detecting HO1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06762. Gene ID: 24451. Target Synonyms: HMOX1; Hsp32; HMOX1D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2710