Learn More
Biomatik Corporation Human Thymic Stromal Lymphopoietin (TSLP) High-sensitive ELISA Kit
Catalog No. 502286966
Encompass_Preferred
Human Thymic Stromal Lymphopoietin (TSLP) High-sensitive ELISA Kit for detecting TSLP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q969D9. Gene ID: 85480. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-228-6966 Supplier Biomatik Corporation Supplier No. EKU11596
Human Thymic Stromal Lymphopoietin (TSLP) High-sensitive ELISA Kit for detecting TSLP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q969D9. Gene ID: 85480. Restrictions: For Research Use Only. Not for use in diagnostic procedures.