Learn More
Biomatik Corporation Mouse Hemojuvelin (HJV) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04726
Encompass_Preferred
Mouse Hemojuvelin (HJV) ELISA Kit for detecting HJV in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.256ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7TQ32. Gene ID: 69585. Target Synonyms: HFE2; HFE2A; HJV; JH; RGMC; hemochromatosis type 2 (Juvenile) ; Haemojuvelin; RGM domain family member C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2712