Learn More
Biomatik Corporation Mouse Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) ELISA Kit, 96 tests
Catalog No. 501502719
Encompass_Preferred
Mouse Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) ELISA Kit for detecting HBEGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06186. Gene ID: 15200. Target Synonyms: HB-EGF; DTR; DTS; DTSF; HEGFL; Diphtheria Toxin Receptor; Proheparin-binding EGF-like growth factor; Diphtheria toxin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2719 Supplier Biomatik Corporation Supplier No. EKU04741
Mouse Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) ELISA Kit for detecting HBEGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06186. Gene ID: 15200. Target Synonyms: HB-EGF; DTR; DTS; DTSF; HEGFL; Diphtheria Toxin Receptor; Proheparin-binding EGF-like growth factor; Diphtheria toxin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.