Learn More
Biomatik Corporation Human Perinuclear Anti-Neutrophil Cytoplasmic Antibody (pANCA) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12108
Encompass_Preferred
Human Perinuclear Anti-Neutrophil Cytoplasmic Antibody (pANCA) Anti-antigen ELISA Kit for detecting pANCA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7071