Learn More
Biomatik Corporation Dog Hepatocyte Growth Factor (HGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04750
Encompass_Preferred
The Dog Hepatocyte Growth Factor (HGF) ELISA Kit allows for quantitative determination of Hepatocyte Growth Factor in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q867B7. Cross Activity: No significant cross-reactivity or interference between Hepatocyte Growth Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2722