Learn More
Biomatik Corporation Ampicillin (ABPC) ELISA Kit
Supplier:  Biomatik Corporation EKU12252
Encompass_Preferred
Ampicillin (ABPC) ELISA Kit for detecting ABPC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 617.3-50,000pg/ml. Sensitivity: 245.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Principen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7102
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'