Learn More
Biomatik Corporation Bovine Cortisol Elisa Kit
Supplier:  Biomatik Corporation EKC31009
Encompass_Preferred
Bovine Cortisol Elisa Kit for detecting Cortisol in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.049-200ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7137