Learn More
Biomatik Corporation Goat Cortisol Elisa Kit
Supplier:  Biomatik Corporation EKC31080
Encompass_Preferred
Goat Cortisol Elisa Kit for detecting Cortisol in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 0.05-200ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7150