Learn More
Biomatik Corporation Human Hepatocyte Growth Factor Activator (HGFAC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04755
Encompass_Preferred
The Human Hepatocyte Growth Factor Activator (HGFAC) ELISA Kit allows for quantitative determination of Hepatocyte Growth Factor Activator in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q04756. Cross Activity: No significant cross-reactivity or interference between Hepatocyte Growth Factor Activator and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2727