Learn More
Biomatik Corporation Pig Hepcidin (Hepc) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04765
Encompass_Preferred
Pig Hepcidin (Hepc) ELISA Kit for detecting Hepc in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 740.74-60000pg/ml. Sensitivity: 283.51pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8MJ80. Gene ID: 397207. Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2730