Learn More
Biomatik Corporation Mouse Hexosaminidase A Alpha (HEXa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04775
Encompass_Preferred
Mouse Hexosaminidase A Alpha (HEXa) ELISA Kit for detecting HEXa in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29416. Gene ID: 15211. Target Synonyms: TSD; Hex-A; Tay Sachs Disease; GM2 Gangliosidosis; Beta-Hexosaminidase Subunit Alpha; Beta-N-Acetylhexosaminidase Subunit Alpha; N-Acetyl-Beta-Glucosaminidase Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2732