Learn More
Biomatik Corporation Human Hexosaminidase B Beta (HEXb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04776
Encompass_Preferred
The Human Hexosaminidase B Beta (HEXb) ELISA Kit allows for quantitative determination of Hexosaminidase B Beta in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07686. Cross Activity: No significant cross-reactivity or interference between Hexosaminidase B Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2733