Learn More
Biomatik Corporation Human High Mobility Group AT Hook Protein 2 (HMGA2) ELISA Kit, 96 tests
Catalog No. 501502736
Encompass_Preferred
The Human High Mobility Group AT Hook Protein 2 (HMGA2) ELISA Kit allows for quantitative determination of High Mobility Group AT Hook Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P52926. Cross Activity: No significant cross-reactivity or interference between High Mobility Group AT Hook Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2736 Supplier Biomatik Corporation Supplier No. EKU04786
The Human High Mobility Group AT Hook Protein 2 (HMGA2) ELISA Kit allows for quantitative determination of High Mobility Group AT Hook Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P52926. Cross Activity: No significant cross-reactivity or interference between High Mobility Group AT Hook Protein 2 and analogues was observed. Restriction: For Research Use Only.