Learn More
Biomatik Corporation Human High Mobility Group Protein 1 (HMG1) ELISA Kit, 96 tests
Catalog No. 501502738
Encompass_Preferred
The Human High Mobility Group Protein 1 (HMG1) ELISA Kit allows for quantitative determination of High Mobility Group Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.61 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09429. Cross Activity: No significant cross-reactivity or interference between High Mobility Group Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2738 Supplier Biomatik Corporation Supplier No. EKU04789
The Human High Mobility Group Protein 1 (HMG1) ELISA Kit allows for quantitative determination of High Mobility Group Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.61 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09429. Cross Activity: No significant cross-reactivity or interference between High Mobility Group Protein 1 and analogues was observed. Restriction: For Research Use Only.