Learn More
Biomatik Corporation Chicken Insulin-Like Growth Factors 1, Igf-1 Elisa Kit
Supplier:  Biomatik Corporation EKC31945
Encompass_Preferred
Chicken Insulin-Like Growth Factors 1, Igf-1 Elisa Kit for detecting Insulin-Like Growth Factor 1 (Somatomedin C) in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 125-2000pg/ml. Sensitivity: 125pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P18254; IGF1. Target Synonyms: IGF1; Insulin-like growth factor I; IGF-I; Somatomedin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7244