Learn More
Biomatik Corporation Mouse High Mobility Group Protein 1 (HMG1) ELISA Kit, 96 tests
Catalog No. 501502739
Encompass_Preferred
Mouse High Mobility Group Protein 1 (HMGB1) ELISA Kit for detecting HMGB1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.88-3,000pg/ml. Sensitivity: 18.29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63158. Gene ID: 15289. Target Synonyms: HMG1; HMG3; SBP1; Sulfoglucuronyl Carbohydrate Binding Protein; Amphoterin; High Mobility Group Box 1 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2739 Supplier Biomatik Corporation Supplier No. EKU04790
Mouse High Mobility Group Protein 1 (HMGB1) ELISA Kit for detecting HMGB1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.88-3,000pg/ml. Sensitivity: 18.29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63158. Gene ID: 15289. Target Synonyms: HMG1; HMG3; SBP1; Sulfoglucuronyl Carbohydrate Binding Protein; Amphoterin; High Mobility Group Box 1 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.