Learn More
Biomatik Corporation Rat High Mobility Group Protein 1 (HMG1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04792
Encompass_Preferred
Rat High Mobility Group Protein 1 (HMGB1) ELISA Kit for detecting HMGB1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63159. Gene ID: 25459. Target Synonyms: HMG1; HMG3; SBP1; Sulfoglucuronyl Carbohydrate Binding Protein; Amphoterin; High Mobility Group Box 1 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2740