Learn More
Biomatik Corporation Human High Molecular Weight Kininogen (HMWK) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04793
Encompass_Preferred
The Human High Molecular Weight Kininogen (HMWK) ELISA Kit allows for quantitative determination of High Molecular Weight Kininogen in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01042. Cross Activity: No significant cross-reactivity or interference between High Molecular Weight Kininogen and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2741