Learn More
Biomatik Corporation Dog Complement 3 (C3) Elisa Kit
Supplier:  Biomatik Corporation EKC32028
Encompass_Preferred
Dog Complement 3 (C3) Elisa Kit for detecting Complement 3(C3) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.78-50ug/ml. Sensitivity: 0.195ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Target Synonyms: AHUS5; ARMD9; ASP; CPAMD1; acylation-stimulating protein cleavage product; complement component C3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7251