Learn More
Biomatik Corporation Histamine (HA) ELISA Kit, 96 tests
Catalog No. 501502742
Encompass_Preferred
The Histamine (HA) ELISA Kit allows for quantitative determination of Histamine in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 1.23-100 ng/mL. Sensitivity: 0.52 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Histamine and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2742 Supplier Biomatik Corporation Supplier No. EKU04794
The Histamine (HA) ELISA Kit allows for quantitative determination of Histamine in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 1.23-100 ng/mL. Sensitivity: 0.52 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Histamine and analogues was observed. Restriction: For Research Use Only.