Learn More
Biomatik Corporation Fish Acetylcholinesterase (Ache) Elisa Kit
Supplier:  Biomatik Corporation EKC32139
Encompass_Preferred
Fish Acetylcholinesterase (Ache) Elisa Kit for detecting Acetylcholinesterase(Ache) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 2.5-40ng/ml. Sensitivity: 2.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P04058; ache. Target Synonyms: acheAcetylcholinesterase; AChE; EC 3.1.1.7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7258