Learn More
Biomatik Corporation Fish Gonadotropin-Releasing Hormone, Gnrh Elisa Kit
Supplier:  Biomatik Corporation EKC32151
Encompass_Preferred
Fish Gonadotropin-Releasing Hormone, Gnrh Elisa Kit for detecting Gonadotropin-Releasing Hormone, Gnrh in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Fish (Vertebrata). Detection Range: 0.5-20ng/ml. Sensitivity: 0.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q5Y835. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7264