Learn More
Biomatik Corporation Fish Heat Shock Protein 70 (Hsp-70) Elisa Kit
Supplier:  Biomatik Corporation EKC32153
Encompass_Preferred
Fish Heat Shock Protein 70 (Hsp-70) Elisa Kit for detecting Heat Shock Protein 70(Hsp-70) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 18.75-300pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q9I8F9. Target Synonyms: Heat shock 70 kDa protein 1; HSP70-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7266