Learn More
Biomatik Corporation Human Histidine Rich Glycoprotein (HRG) ELISA Kit, 96 tests
Catalog No. 501502747
Encompass_Preferred
The Human Histidine Rich Glycoprotein (HRG) ELISA Kit allows for quantitative determination of Histidine Rich Glycoprotein in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47-30 ng/mL. Sensitivity: 0.22 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04196. Cross Activity: No significant cross-reactivity or interference between Histidine Rich Glycoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2747 Supplier Biomatik Corporation Supplier No. EKU04805
The Human Histidine Rich Glycoprotein (HRG) ELISA Kit allows for quantitative determination of Histidine Rich Glycoprotein in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47-30 ng/mL. Sensitivity: 0.22 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04196. Cross Activity: No significant cross-reactivity or interference between Histidine Rich Glycoprotein and analogues was observed. Restriction: For Research Use Only.