Learn More
Biomatik Corporation Fish Interleukin 6, Il-6 Elisa Kit
Supplier:  Biomatik Corporation EKC32161
Encompass_Preferred
Fish Interleukin 6, Il-6 Elisa Kit for detecting Interleukin 6, Il-6 in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 12.5-200pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: A0S0B0; il6. Target Synonyms: il6Interleukin-6; IL-6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7272