Learn More
Biomatik Corporation Fish Tumor Necrosis Factor Α (Tnf-Α) Elisa Kit
Supplier:  Biomatik Corporation EKC32171
Encompass_Preferred
Fish Tumor Necrosis Factor Α (Tnf-Α) Elisa Kit for detecting Tumor Necrosis Factor Α(Tnf-Α) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 125-5000pg/ml. Sensitivity: 125pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q8JFG3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7277