Learn More
Biomatik Corporation Human Histone Deacetylase 6 (HDAC6) ELISA Kit, 96 tests
Catalog No. 501502750
Encompass_Preferred
The Human Histone Deacetylase 6 (HDAC6) ELISA Kit allows for quantitative determination of Histone Deacetylase 6 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UBN7. Cross Activity: No significant cross-reactivity or interference between Histone Deacetylase 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2750 Supplier Biomatik Corporation Supplier No. EKU04819
The Human Histone Deacetylase 6 (HDAC6) ELISA Kit allows for quantitative determination of Histone Deacetylase 6 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UBN7. Cross Activity: No significant cross-reactivity or interference between Histone Deacetylase 6 and analogues was observed. Restriction: For Research Use Only.