Learn More
Biomatik Corporation Mouse Histone Deacetylase 6 (HDAC6) ELISA Kit, 96 tests
Catalog No. 501502751
Encompass_Preferred
Mouse Histone Deacetylase 6 (HDAC6) ELISA Kit for detecting HDAC6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2V5. Gene ID: 15185. Target Synonyms: HD6; JM21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2751 Supplier Biomatik Corporation Supplier No. EKU04820
Mouse Histone Deacetylase 6 (HDAC6) ELISA Kit for detecting HDAC6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2V5. Gene ID: 15185. Target Synonyms: HD6; JM21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.