Learn More
Biomatik Corporation Mouse Homing Associated Cell Adhesion Molecule (HCAM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04832
Encompass_Preferred
Mouse Homing Associated Cell Adhesion Molecule (HCAM) ELISA Kit for detecting HCAM in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15379. Gene ID: 12505. Target Synonyms: CDW44; CD44; CSPG8; ECMR-III; HCELL; IN; LHR; MC56; MDU2; MDU3; MIC4; MUTCH-I; Pgp1; Hematopoietic Cell E- And L-Selectin Ligand; Chondroitin Sulfate Proteoglycan 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2752