Learn More
Biomatik Corporation Grouper Vitellogenin, Vtg Elisa Kit
Supplier:  Biomatik Corporation EKC32240
Encompass_Preferred
Grouper Vitellogenin, Vtg Elisa Kit for detecting Vitellogenin, Vtg in serum, plasma, tissue homogenates. Species Reactivity: Grouper. Detection Range: 50-2000ng/ml. Sensitivity: 50ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q92093; vtg1. Target Synonyms: vtg1; Vitellogenin; VTG) [Cleaved into: Lipovitellin I; LVI); Phosvitin; PV); Lipovitellin II; LVII)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7286