Learn More
Biomatik Corporation Human High Temperature Requirement Factor A1 (HTRA1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04837
Encompass_Preferred
The Human High Temperature Requirement Factor A1 (HTRA1) ELISA Kit allows for quantitative determination of High Temperature Requirement Factor A1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92743. Cross Activity: No significant cross-reactivity or interference between High Temperature Requirement Factor A1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2754