Learn More
Biomatik Corporation Human 8-Hydroxydeoxyguanosine (8-OHdG) ELISA kit
Supplier:  Biomatik Corporation EKC32422
Encompass_Preferred
Sample Type: serum, urine. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12 ng/mL-800 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7298