Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 12,ADAM12 ELISA Kit
Supplier:  Biomatik Corporation EKC32426
Encompass_Preferred
Sample Type: serum, plasma, urine, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O43184. Target Synonyms: RP11-295J3.5, MCMP, MCMPMltna, MLTN, MLTNA, a disintegrin and metalloproteinase domain 12 (meltrin alpha)|meltrin alpha. Restriction: For Research Use Only.
Catalog No. 50-228-7300