Learn More
Biomatik Corporation Human Adamts-Like Protein 2 (Adamtsl2) Elisa Kit
Supplier:  Biomatik Corporation EKC32488
Encompass_Preferred
Human Adamts-Like Protein 2 (Adamtsl2) Elisa Kit for detecting Adamts-Like 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86TH1; ADAMTSL2. Target Synonyms: ADAMTS like 2; ADAMTS like protein 2; ADAMTS-like protein 2; ADAMTSL 2; ADAMTSL-2; ADAMTSL2; ATL2_HUMAN; FLJ45164; KIAA0605. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7306