Learn More
Biomatik Corporation Human Aggrecan Core Protein (Agc1) Elisa Kit
Supplier:  Biomatik Corporation EKC32511
Encompass_Preferred
Human Aggrecan Core Protein (Agc1) Elisa Kit for detecting Aggrecan 1 (Chondroitin Sulfate Proteoglycan 1, Large Aggregating Proteoglycan, Antigen Identified By Monoclonal Antibody A0122) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16112; ACAN. Target Synonyms: ACAN; AGC1; CSPG1; MSK16; Aggrecan core protein; Cartilage-specific proteoglycan core protein; CSPCP; Chondroitin sulfate proteoglycan core protein 1; Chondroitin sulfate proteoglycan 1) [Cleaved into: Aggrecan core protein 2]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7308